Learn More
Aviva Systems Biology Corp CY24B Antibody - C-terminal region (ARP44276_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44276_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYBB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CY24B. Alternate names: CGD, NOX2, IMD34, AMCBX2, GP91-1, GP91PHOX, p91-PHOX, GP91-PHOX, ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53413