Learn More
Enterprise Technology Solutions CyberPower BP72V60ART2U Extended Battery Module
SDP
Supplier:  Enterprise Technology Solutions BP72V60ART2U
Encompass_Preferred
The BP72V60ART2U Extended Battery Module from CyberPower will extend the battery life of select models of their Smart App Online UPS systems during power outages. This battery module features a hot-swappable and user-replaceable battery pack consisting of twelve 12V/9Ah sealed lead-acid cells with a typical recharge time of 4 hours. Thanks to a 2 RU form factor, this module can be installed in a rack or tower configuration to match the form factor of the UPS it is supporting.
Catalog No. 50-001-73777