Learn More
Enterprise Technology Solutions CyberPower M750L Medical-Grade UPS
SDP
Catalog No. 5000173578
Encompass_Preferred
The M750L Medical-Grade UPS from CyberPower delivers clean, consistent backup power to sensitive medical equipment in hospitals and healthcare facilities. Designed for use in patient care environments, the M750L has been tested to meet UL 60601-1 requirements and includes hospital-grade plugs and outlets. It also features a built-in isolation transformer to help minimize current leakage and eliminate line noise,
Catalog No. 50-001-73578 Supplier Enterprise Technology Solutions Supplier No. M750L
The M750L Medical-Grade UPS from CyberPower delivers clean, consistent backup power to sensitive medical equipment in hospitals and healthcare facilities. Designed for use in patient care environments, the M750L has been tested to meet UL 60601-1 requirements and includes hospital-grade plugs and outlets. It also features a built-in isolation transformer to help minimize current leakage and eliminate line noise,