Learn More
Enterprise Technology Solutions CyberPower PR1000ELCDRT2U Smart App Sinewave UPS (1000VA/900W)
SDP
Catalog No. 5000172454
Encompass_Preferred
The CyberPower PR1000ELCDRT2U Smart App Sinewave UPS (1000VA/900W) is a convertible rack/tower UPS with line interactive topology and an adjustable output of 220, 230, or 240VAC. It provides battery backup, power protection with sine wave output, and surge protection for corporate servers,
Catalog No. 50-001-72454 Supplier Enterprise Technology Solutions Supplier No. PR1000ELCDRT2U
The CyberPower PR1000ELCDRT2U Smart App Sinewave UPS (1000VA/900W) is a convertible rack/tower UPS with line interactive topology and an adjustable output of 220, 230, or 240VAC. It provides battery backup, power protection with sine wave output, and surge protection for corporate servers,