Learn More
Enterprise Technology Solutions CyberPower PR1000RT2UC Smart App Sinewave UPS
SDP
Catalog No. 5000172543
Encompass_Preferred
Keep your hardware protected from spikes and outages with the PR1000RT2UC Smart App Sinewave UPS from CyberPower, a rack/tower convertible UPS with line-interactive topology, surge protection, battery backup, and sine wave output to eliminate power gaps that can be detected by sensitive equipment and cause unexpected shutdowns.
Catalog No. 50-001-72543 Supplier Enterprise Technology Solutions Supplier No. PR1000RT2UC
Keep your hardware protected from spikes and outages with the PR1000RT2UC Smart App Sinewave UPS from CyberPower, a rack/tower convertible UPS with line-interactive topology, surge protection, battery backup, and sine wave output to eliminate power gaps that can be detected by sensitive equipment and cause unexpected shutdowns.