Learn More
Enterprise Technology Solutions CyberPower RB1270X4B UPS Replacement Battery Cartridges
SDP
Catalog No. 5000172511
Encompass_Preferred
Designed for their OR1500LCDRTXL2U UPS system, the RB1270X4B UPS Replacement Battery Cartridges from CyberPower contains four leakproof and sealed lead-acid batteries with a capacity of 12V / 7Ah. They are OEM-certified to the manufacturing standards of your UPS' original battery, its pre-assembled cartridges can be installed by the user.
Catalog No. 50-001-72511 Supplier Enterprise Technology Solutions Supplier No. RB1270X4B
Designed for their OR1500LCDRTXL2U UPS system, the RB1270X4B UPS Replacement Battery Cartridges from CyberPower contains four leakproof and sealed lead-acid batteries with a capacity of 12V / 7Ah. They are OEM-certified to the manufacturing standards of your UPS' original battery, its pre-assembled cartridges can be installed by the user.