Learn More
Enterprise Technology Solutions CyberPower RB1280X2A UPS Battery Cartridges
SDP
Catalog No. 5000173453
Encompass_Preferred
Designed for select CyberPower UPS systems, the RB1280X2A UPS Battery Cartridges from CyberPower contain leakproof and sealed lead acid batteries with a capacity of 12V, 8Ah. They are OEM certified to the manufacturing standards of your UPS' original battery.
Catalog No. 50-001-73453 Supplier Enterprise Technology Solutions Supplier No. RB1280X2A
Designed for select CyberPower UPS systems, the RB1280X2A UPS Battery Cartridges from CyberPower contain leakproof and sealed lead acid batteries with a capacity of 12V, 8Ah. They are OEM certified to the manufacturing standards of your UPS' original battery.