Learn More
Enterprise Technology Solutions CyberPower RB1290X2E UPS Replacement Battery Cartridge
SDP
Catalog No. 5000173168
Encompass_Preferred
The RB1290X2E UPS Replacement Battery Cartridge from CyberPower replaces weak or completely depleted internal batteries within their CP1500AVRLCD3, CP1350AVRLCD3, and BRG1500AVRLCD2 UPS systems. This replacement is comprised of two 12V/9Ah batteries that operate using sealed lead-acid chemistry and leak-proof casing.
Catalog No. 50-001-73168 Supplier Enterprise Technology Solutions Supplier No. XER006R04383
The RB1290X2E UPS Replacement Battery Cartridge from CyberPower replaces weak or completely depleted internal batteries within their CP1500AVRLCD3, CP1350AVRLCD3, and BRG1500AVRLCD2 UPS systems. This replacement is comprised of two 12V/9Ah batteries that operate using sealed lead-acid chemistry and leak-proof casing.