Learn More
Enterprise Technology Solutions CyberPower RB1290X3R UPS Replacement Battery Cartridge
SDP
Catalog No. 5000173366
Encompass_Preferred
Designed for their BP36V60ART2U Extended Battery Module (EMB), the RB1290X3R UPS Replacement Battery Cartridge from CyberPower contains three leakproof and sealed lead-acid batteries with a capacity of 12V / 9Ah. They are OEM-certified to the manufacturing standards of your UPS' original battery.
Catalog No. 50-001-73366 Supplier Enterprise Technology Solutions Supplier No. RB1290X3R
Designed for their BP36V60ART2U Extended Battery Module (EMB), the RB1290X3R UPS Replacement Battery Cartridge from CyberPower contains three leakproof and sealed lead-acid batteries with a capacity of 12V / 9Ah. They are OEM-certified to the manufacturing standards of your UPS' original battery.