Learn More
ANALYTIK JENA US LLC Analytik Jena CyBio FeliX Basic Unit, Automated Liquid Handling, Modular Design, 12 Deck Positions
Catalog No. NC3734410
Encompass
The Analytik Jena CyBio FeliX Basic Unit is an automated liquid handling platform designed for precise, medium to high-throughput applications in laboratory settings. It features a compact design with twelve deck positions on two levels, allowing efficient processing of various well formats and microtubes. Its modular and adaptable configuration, with interchangeable pipetting heads and safety features, enhances productivity while maintaining a clean working environment.
Catalog No. NC3734410 Supplier ANALYTIK JENA US LLC Supplier No. OL501525500
Add to Cart
Add to Cart
Request a Quote
The Analytik Jena CyBio FeliX Basic Unit is an automated liquid handling platform designed for precise, medium to high-throughput applications in laboratory settings. It features a compact design with twelve deck positions on two levels, allowing efficient processing of various well formats and microtubes. Its modular and adaptable configuration, with interchangeable pipetting heads and safety features, enhances productivity while maintaining a clean working environment.