Learn More
ANALYTIK JENA US LLC CyBio FeliX Head R 96 Channels, High Precision, Positive Air Displacement
Supplier:  ANALYTIK JENA US LLC OL331614850
Encompass
The CyBio FeliX Head R 96/250 µL is a high-precision pipetting head designed for the CyBio FeliX liquid handling platform. It features 96 channels and delivers accurate liquid transfers up to 250 µL using positive air displacement technology. Its modular design allows for customizable performance, enhancing efficiency in various laboratory applications such as high-throughput screening and assay development. Built with quality materials, it ensures long-lasting reliability with minimal maintenance.
Catalog No. NC3734412
Only null left
Add to Cart
Request a Quote