Learn More
ANALYTIK JENA US LLC CyBio TipTray 384/10 µL, Disposable Tips, Compatible With CyBio Liquid Handlers, 1 Tray
Catalog No. NC2294551
Encompass
The CyBio TipTray 384/10 µL from Analytik Jena is a high-precision disposable tip tray designed for 384-well microplate formats. It ensures accurate liquid handling in automated systems, enhancing high-throughput screening and replication tasks. With 384 tips per tray and proprietary sealing technology, it guarantees repeatable results in various applications, making it a reliable choice for scientific laboratories.
Catalog No. NC2294551 Supplier ANALYTIK JENA US LLC Supplier No. OL380025512N
The CyBio TipTray 384/10 µL from Analytik Jena is a high-precision disposable tip tray designed for 384-well microplate formats. It ensures accurate liquid handling in automated systems, enhancing high-throughput screening and replication tasks. With 384 tips per tray and proprietary sealing technology, it guarantees repeatable results in various applications, making it a reliable choice for scientific laboratories.