Learn More
Ellsworth Adhesive Co Cyberbond 6020 Cyanoacrylate Adhesive Clear 2 Oz Bottle
Supplier:  Ellsworth Adhesive Co 6020 2 OZ BOTTLE
Encompass
This item has a minimum qty of 20 per supplier requirements. HB Fuller Cyberbond 6020 Cyanoacrylate Adhesive Clear is used in the removal from metal, plastic, and rubber surfaces. It quickly softens and dissolves adhesive from unwanted areas. 2 oz Bottle.
Catalog No. NC1701826