Learn More
Medchemexpress LLC CYC-116 10mg | 693228-63-6 | 10MG
SDP
Catalog No. 501968276
Encompass_Preferred
CYC-116 is a potent aurora A and aurora B inhibitor with Kis of 8 and 9 nM respectively
Catalog No. 50-196-8276 Supplier Medchemexpress LLC Supplier No. HY1055810MG
CYC-116 is a potent aurora A and aurora B inhibitor with Kis of 8 and 9 nM respectively