Learn More
TARGETMOL CHEMICALS INC CYC-116 25MG
SDP
Catalog No. 502579357
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CYC116 is a potent inhibitor of Aurora A/B with Ki of 8.0 nM/9.2 nM is less potent to VEGFR2 (Ki of 44 nM) with 50-fold greater potency than CDKs not active against PKA Akt/PKB PKC no effect on GSK-3(alpha)/(beta) CK2 Plk1 and SAPK2A. Phase 1. purity: 96%
Catalog No. 50-257-9357 Supplier TARGETMOL CHEMICALS INC Supplier No. T645825MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CYC116 is a potent inhibitor of Aurora A/B with Ki of 8.0 nM/9.2 nM is less potent to VEGFR2 (Ki of 44 nM) with 50-fold greater potency than CDKs not active against PKA Akt/PKB PKC no effect on GSK-3(alpha)/(beta) CK2 Plk1 and SAPK2A. Phase 1. purity: 96%