Learn More
Aviva Systems Biology Corp Cyc1 Antibody - C-terminal region (ARP51993_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51993_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cyc1. It was validated on Western Blot. . Alternate names: 2610002H19Rik, AA408921ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-620