Learn More
Selleck Chemical LLC CYC116 10mg 693228-63-6
SDP
Catalog No. 506888
Encompass_Preferred
CYC116 is a potent inhibitor of Aurora A/B with Ki of 8.0 nM/9.2 nM, is less potent to VEGFR2 (Ki of 44 nM), with 50-fold greater potency than CDKs, not active against PKA, Akt/PKB, PKC, no effect on GSK-3?/?, CK2, Plk1 and SAPK2A. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-688-8 Supplier Selleck Chemical LLC Supplier No. S117110MG
CYC116 is a potent inhibitor of Aurora A/B with Ki of 8.0 nM/9.2 nM, is less potent to VEGFR2 (Ki of 44 nM), with 50-fold greater potency than CDKs, not active against PKA, Akt/PKB, PKC, no effect on GSK-3?/?, CK2, Plk1 and SAPK2A. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.