Learn More
Medchemexpress LLC Cyclamic Acid-d11 | 1215770-73-2 | 99.9% | 1 MG
SDP
Catalog No. 5000405489
Encompass_Preferred
Cyclamic Acid-d11 is the deuterium labeled Cyclamic acid, a widely used artificial sweetener in food and pharmaceuticals. This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes like deuterium have been incorporated into drug molecules for quantitation during drug development
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-05489 Supplier Medchemexpress LLC Supplier No. HYB0541S1MG
Cyclamic Acid-d11 is the deuterium labeled Cyclamic acid, a widely used artificial sweetener in food and pharmaceuticals. This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes like deuterium have been incorporated into drug molecules for quantitation during drug development
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs