Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclamic acid (sodium) | 139-05-9 | MFCD00003827 | 98.0% | 201.22 | C6H12NNaO3S | 5 G
SDP

Catalog No. 5000303272
Encompass_Preferred

Cyclamic acid (sodium), commonly known as sodium cyclamate (CAS 139-05-9), is the sodium salt of cyclamic acid used as an analytical reference standard and research reagent. Supplied as a white crystalline solid, it is used in sweetener research, biochemical assays, and analytical method development. Intended for research use only.

  • High purity (98.0%).
  • Molecular weight 201.22 g/mol.
  • Chemical formula C6H12NNaO3S.
  • White crystalline powder, water soluble.
  • Suitable for analytical and biochemical applications.
  • Available in small solid packs for laboratory use.

Catalog No. 50-003-03272 Supplier Medchemexpress LLC Supplier No. HYW0148395G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclamic acid (sodium), commonly known as sodium cyclamate (CAS 139-05-9), is the sodium salt of cyclamic acid used as an analytical reference standard and research reagent. Supplied as a white crystalline solid, it is used in sweetener research, biochemical assays, and analytical method development. Intended for research use only.

  • High purity (98.0%).
  • Molecular weight 201.22 g/mol.
  • Chemical formula C6H12NNaO3S.
  • White crystalline powder, water soluble.
  • Suitable for analytical and biochemical applications.
  • Available in small solid packs for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.