Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclamic acid (sodium) | 139-05-9 | MFCD00003827 | 99.9% | 201.22 g/mol | C6H12NNaO3S | 1 ML
SDP

Catalog No. 5000303271
Encompass_Preferred

Cyclamic acid (sodium) is the sodium salt of cyclohexylsulfamic acid supplied for research and analytical applications in solid and pre-made solution formats. It is used as an artificial sweetener reference and an analytical standard for chromatographic and mass spectrometry methods.

  • High purity: 99.94% (analytical standard)
  • Available as 10 mM x 1 mL solution in DMSO for convenience
  • Suitable for HPLC, GC, and mass spectrometry workflows
  • Solubility guidance provided for water and DMSO to aid assay preparation
  • Storage recommendations: solid at 4°C; in solvent at -80°C (6 months) or -20°C (1 month)

Catalog No. 50-003-03271 Supplier Medchemexpress LLC Supplier No. HYW01483910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclamic acid (sodium) is the sodium salt of cyclohexylsulfamic acid supplied for research and analytical applications in solid and pre-made solution formats. It is used as an artificial sweetener reference and an analytical standard for chromatographic and mass spectrometry methods.

  • High purity: 99.94% (analytical standard)
  • Available as 10 mM x 1 mL solution in DMSO for convenience
  • Suitable for HPLC, GC, and mass spectrometry workflows
  • Solubility guidance provided for water and DMSO to aid assay preparation
  • Storage recommendations: solid at 4°C; in solvent at -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.