Learn More
Medchemexpress LLC Cyclanilide | 113136-77-9 | MFCD03792791 | 99.9% | 274.10 g·mol⁻¹ | C11H9Cl2NO3 | 250 MG
SDP
Catalog No. 5000227329
Encompass_Preferred
Cyclanilide is a plant growth regulator used in research to influence cotton growth stages and auxin-regulated processes. Supplied as an analytical reagent for laboratory use only.
  • Plant growth regulator applied to cotton during various growth stages.
  • High purity analytical reagent (99.9%).
  • Solid, off-white to light-yellow physical form.
  • Molecular formula C11H9Cl2NO3 with molecular weight 274.10 g·mol⁻¹.
  • Intended for laboratory and research use only.
Catalog No. 50-002-27329 Supplier Medchemexpress LLC Supplier No. HY118217250MG
Cyclanilide is a plant growth regulator used in research to influence cotton growth stages and auxin-regulated processes. Supplied as an analytical reagent for laboratory use only.
  • Plant growth regulator applied to cotton during various growth stages.
  • High purity analytical reagent (99.9%).
  • Solid, off-white to light-yellow physical form.
  • Molecular formula C11H9Cl2NO3 with molecular weight 274.10 g·mol⁻¹.
  • Intended for laboratory and research use only.