Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Life Cycles of Plants and Animals Learning Guide
SCI_ED

Catalog No. s13894 Shop All NewPath Learning Products
Change view
Click to view available options
Format:
36-page workbook
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S13894 36-page workbook
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13894 Supplier NewPath Learning Supplier No. 143261
Only null left
Add to Cart
Add to Cart

Use engaging graphics and inquiry-based activities to enhance student comprehension on standards-based concepts on Life Cycles

Full-color, 36-page guide features self-directed readings, easy-to-follow illustrated explanations; guiding questions; inquiry-based activities; a lab investigation; key vocabulary review; and assessment review questions along with a post-test

Topics covered:

  • Plant Parts
  • Bean or Radish Life Cycle
  • Frog
  • Insect (Butterfly)
  • Inheritance of Traits
  • Animal Variation
  • Adaptation

Specifications

Product Type Learning Guide
Format 36-page workbook
For Use With (Application) Enhance student comprehension on standards-based concepts on life cycles
Certifications/Compliance Meets Academic Standard for California, Florida, National, New York, Texas and Next Generation Science Standards
Quantity 10/Pk.
Grade Level Elementary
Grade 3 to 5
Subject Life Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.