Learn More
PURSOLUTIONS LLC Cycled Tubulin, 5mg (5x1 mg)
Catalog No. NC2754239
Encompass
Purity: >99%Source: bovineMolecular Weight: ~110 kDaForm: clear aqueous solutionConcentration: 20 mg/mlBuffer Conditions: 80 mM PIPES, 1 mM EGTA, and 1 mM MgCl2 (pH 6.8)Shipping: shipped on dry iceStorage Conditions: store at -80˚C immediatelyShelf Life: check product label for expiration dateFor research use only. Not for medical or diagnostic use.
Catalog No. NC2754239 Supplier PURSOLUTIONS LLC Supplier No. 0320055X1MG
Purity: >99%Source: bovineMolecular Weight: ~110 kDaForm: clear aqueous solutionConcentration: 20 mg/mlBuffer Conditions: 80 mM PIPES, 1 mM EGTA, and 1 mM MgCl2 (pH 6.8)Shipping: shipped on dry iceStorage Conditions: store at -80˚C immediatelyShelf Life: check product label for expiration dateFor research use only. Not for medical or diagnostic use.