Learn More
Apexbio Technology LLC CYCLEN-200MG
SDP
Catalog No. 5000568632
Encompass_Preferred
Cyclen (CAS No 294-90-6) is a significant compound in the field of biomedical research exploring its role in coordination chemistry and its potential applications in drug development Originating from synthetic chemical processes Cyclen is a macrocyclic ligand that primarily targets metal ions influencing metalloprotein interactions Its mechanism involves binding to metal ions and forming stable complexes which is crucial in modulating biological metal ion pathways In vitro studies often demonstrate its activity in the nanomolar to low micromolar range underscoring its effectiveness in high-affinity binding scenarios Cyclen is frequently utilized in various research applications including studies involving cell models and animal experiments and holds potential in drug screening for therapeutic development Experimental concentrations of Cyclen typically depend on the specific research design ensuring optimized conditions are met for desired outcomes
Catalog No. 50-005-68632 Supplier Apexbio Technology LLC Supplier No. C6632200
Cyclen (CAS No 294-90-6) is a significant compound in the field of biomedical research exploring its role in coordination chemistry and its potential applications in drug development Originating from synthetic chemical processes Cyclen is a macrocyclic ligand that primarily targets metal ions influencing metalloprotein interactions Its mechanism involves binding to metal ions and forming stable complexes which is crucial in modulating biological metal ion pathways In vitro studies often demonstrate its activity in the nanomolar to low micromolar range underscoring its effectiveness in high-affinity binding scenarios Cyclen is frequently utilized in various research applications including studies involving cell models and animal experiments and holds potential in drug screening for therapeutic development Experimental concentrations of Cyclen typically depend on the specific research design ensuring optimized conditions are met for desired outcomes