Learn More
Medchemexpress LLC Cyclen | 294-90-6 | 172.28 | 50 G
SDP
Catalog No. 5000445649
Encompass_Preferred
Cyclen is a macrocyclic tetraamine chelating agent and an aza analogue of crown ether. It serves as a precursor for MRI contrast agents and an intermediate for effective macrocyclic chelates. It is also used as a structural regulator in interfacial polymerization, developing polyamide NF membranes for efficient lithium-magnesium separation due to its selective coordination properties for lithium ions.
  • Macrocyclic tetraamine chelating agent
  • Precursor for MRI contrast agents
  • Intermediate for preparing effective macrocyclic chelates
  • Regulator in interfacial polymerization for polyamide NF membranes
  • Facilitates efficient lithium-magnesium separation
  • Binds dehydrated lithium with high adsorption energy
  • Forms stable complexes with lithium ions
  • Used in nanofiltration for lithium recovery
Catalog No. 50-004-45649 Supplier Medchemexpress LLC Supplier No. HYW00765650G
Cyclen is a macrocyclic tetraamine chelating agent and an aza analogue of crown ether. It serves as a precursor for MRI contrast agents and an intermediate for effective macrocyclic chelates. It is also used as a structural regulator in interfacial polymerization, developing polyamide NF membranes for efficient lithium-magnesium separation due to its selective coordination properties for lithium ions.
  • Macrocyclic tetraamine chelating agent
  • Precursor for MRI contrast agents
  • Intermediate for preparing effective macrocyclic chelates
  • Regulator in interfacial polymerization for polyamide NF membranes
  • Facilitates efficient lithium-magnesium separation
  • Binds dehydrated lithium with high adsorption energy
  • Forms stable complexes with lithium ions
  • Used in nanofiltration for lithium recovery