Learn More
Medchemexpress LLC Cyclic adenosine monophosphate | 60-92-4 | 99.89% | 50 MG
SDP
Catalog No. 5000411958
Encompass_Preferred
Cyclic AMP (Standard) is the analytical standard of Cyclic AMP. Cyclic AMP (Cyclic adenosine monophosphate), an adenosine triphosphate derivative, is an intracellular signaling molecule responsible for directing cellular responses to extracellular signals. It is an important second messenger in many biological processes.
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications
Catalog No. 50-004-11958 Supplier Medchemexpress LLC Supplier No. HYB1511R50MG
Cyclic AMP (Standard) is the analytical standard of Cyclic AMP. Cyclic AMP (Cyclic adenosine monophosphate), an adenosine triphosphate derivative, is an intracellular signaling molecule responsible for directing cellular responses to extracellular signals. It is an important second messenger in many biological processes.
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications