Learn More
Medchemexpress LLC Cyclic adenosine monophosphate | 60-92-4 | 99.9% | 10 G
SDP
Catalog No. 5000411476
Encompass_Preferred
Cyclic AMP (Cyclic adenosine monophosphate) is a derivative of adenosine triphosphate and functions as an intracellular signaling molecule, directing cellular responses to extracellular signals. It is recognized as an important second messenger in numerous biological processes. Cyclic AMP modulates mediator generation by suppressing the expression of pro-inflammatory cytokines, such as TNF-α and IL-12, while enhancing the production of the anti-inflammatory cytokine IL-10.
  • Intracellular signaling molecule
  • Directs cellular responses to extracellular signals
  • Functions as an important second messenger in biological processes
  • Modulates mediator generation
  • Suppresses pro-inflammatory cytokines
  • Enhances anti-inflammatory cytokine production
Catalog No. 50-004-11476 Supplier Medchemexpress LLC Supplier No. HYB151110G
Cyclic AMP (Cyclic adenosine monophosphate) is a derivative of adenosine triphosphate and functions as an intracellular signaling molecule, directing cellular responses to extracellular signals. It is recognized as an important second messenger in numerous biological processes. Cyclic AMP modulates mediator generation by suppressing the expression of pro-inflammatory cytokines, such as TNF-α and IL-12, while enhancing the production of the anti-inflammatory cytokine IL-10.
  • Intracellular signaling molecule
  • Directs cellular responses to extracellular signals
  • Functions as an important second messenger in biological processes
  • Modulates mediator generation
  • Suppresses pro-inflammatory cytokines
  • Enhances anti-inflammatory cytokine production