Learn More
Medchemexpress LLC Cyclic adenosine monophosphate | 60-92-4 | 99.9% | 100 MG
SDP
Catalog No. 5000411360
Encompass_Preferred
Cyclic AMP (Standard) is the analytical standard of Cyclic AMP, an adenosine triphosphate derivative and an intracellular signaling molecule. It is an important second messenger in many biological processes, directing cellular responses to extracellular signals.
  • Used as a reference standard in assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for applications involving HPLC, GC, and MS
Catalog No. 50-004-11360 Supplier Medchemexpress LLC Supplier No. HYB1511R100MG
Add to Cart
Add to Cart
Cyclic AMP (Standard) is the analytical standard of Cyclic AMP, an adenosine triphosphate derivative and an intracellular signaling molecule. It is an important second messenger in many biological processes, directing cellular responses to extracellular signals.
  • Used as a reference standard in assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for applications involving HPLC, GC, and MS