Learn More
Medchemexpress LLC Cyclic ADP-ribose | 119340-53-3 | 500 UG
SDP
Catalog No. 5000418656
Encompass_Preferred
Cyclic ADP-ribose (cADPR) is a powerful second messenger involved in calcium mobilization. It is synthesized from NAD+ by an enzyme called ADP-ribosyl cyclase. cADPR primarily elevates cytosolic calcium levels through the Ryanodine receptor-mediated release of calcium from the endoplasmic reticulum, and also by facilitating extracellular calcium influx via the opening of TRPM2 channels.
  • Potent second messenger for calcium mobilization
  • Synthesized from NAD+ by ADP-ribosyl cyclase
  • Increases cytosolic calcium via Ryanodine receptor-mediated release
  • Facilitates extracellular calcium influx via TRPM2 channels
  • Endogenous metabolite
  • Purity of ≥96.0%
  • Solid, white to off-white appearance
  • Primarily intended for research purposes
  • Used to study calcium release in various biological systems
  • Investigated for its role in regulating oxytocin release
Catalog No. 50-004-18656 Supplier Medchemexpress LLC Supplier No. HYN7395500UG
Cyclic ADP-ribose (cADPR) is a powerful second messenger involved in calcium mobilization. It is synthesized from NAD+ by an enzyme called ADP-ribosyl cyclase. cADPR primarily elevates cytosolic calcium levels through the Ryanodine receptor-mediated release of calcium from the endoplasmic reticulum, and also by facilitating extracellular calcium influx via the opening of TRPM2 channels.
  • Potent second messenger for calcium mobilization
  • Synthesized from NAD+ by ADP-ribosyl cyclase
  • Increases cytosolic calcium via Ryanodine receptor-mediated release
  • Facilitates extracellular calcium influx via TRPM2 channels
  • Endogenous metabolite
  • Purity of ≥96.0%
  • Solid, white to off-white appearance
  • Primarily intended for research purposes
  • Used to study calcium release in various biological systems
  • Investigated for its role in regulating oxytocin release