Learn More
Cayman Chemical CyclIc ADP-RIboseamonIM500ug
SDP
Catalog No. 502994275
Encompass_Preferred
An endogenous NAD+ metabolite that mobilizes intracellular Ca2+ release via ryanodine receptors; may trigger Ca2+ influx via TRPM2
Catalog No. 50-299-4275 Supplier Cayman Chemical Supplier No. 21417500 UG
An endogenous NAD+ metabolite that mobilizes intracellular Ca2+ release via ryanodine receptors; may trigger Ca2+ influx via TRPM2