Learn More
Medchemexpress LLC HY-N7395A 500ug Medchemexpress, Cyclic ADP-​ribose (ammonium) CAS: Purity:>98%
SDP
Catalog No. 502082995
Encompass_Preferred
Medchemexpress, HY-N7395A 500ug Cyclic ADP-​ribose (ammonium) CAS: Cyclic ADP-ribose ammonium (cADPR ammonium) is a potent second messenger for calcium mobilization that is synthesized from NAD+ by an ADP-ribosyl cyclase. Cyclic ADP-ribose ammonium increases cytosolic calcium mainly by Ryanodine receptor-mediated release from endoplasmic reticulum and also by extracellular influx through the opening of TRPM2 channels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2995 Supplier Medchemexpress LLC Supplier No. HYN7395A500UG
Medchemexpress, HY-N7395A 500ug Cyclic ADP-​ribose (ammonium) CAS: Cyclic ADP-ribose ammonium (cADPR ammonium) is a potent second messenger for calcium mobilization that is synthesized from NAD+ by an ADP-ribosyl cyclase. Cyclic ADP-ribose ammonium increases cytosolic calcium mainly by Ryanodine receptor-mediated release from endoplasmic reticulum and also by extracellular influx through the opening of TRPM2 channels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.