Learn More
Medchemexpress LLC Cyclic AMP-13C5 (Cyclic adenosine monophosphate-13C5) | 2170095-23-3 | 97.5% | 1 MG
SDP
Catalog No. 5000411899
Encompass_Preferred
Cyclic AMP-13C5 is a deuterated cyclic AMP. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes.
Catalog No. 50-004-11899 Supplier Medchemexpress LLC Supplier No. HYB1511S1MG
Cyclic AMP-13C5 is a deuterated cyclic AMP. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes.