Learn More
American Radiolabeled Chemicals Inc CYCLIC AMP 3H 250UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 1790250UC
Encompass_Preferred
Adenosine 3',5'-cyclic monophosphate-5',8-3H ammonium salt 250uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 60-90 Ci/mmol, M.W. 346.30, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-219