Learn More
Cayman Chemical CyclIc AMP AChE Tracer 500dtn
SDP
Catalog No. 502944586
Encompass_Preferred
A steroidal glycoside with diverse biological activities reduces the proliferation of LNCaP and T47D cancer cells (IC50s 32.1 and 27.5 UM respectively) inhibits TNF-a-induced nuclear translocation of NF-KB in HaCaT keratinocytes at 50 nM decreases AST serum levels in a mouse model of carbon tetrachloride-induced liver injury at 0.1 mg/kg topical administration reduces ear epidermal thickness and scaling as well as leukocyte and macrophage infiltration in an imiquimod-induced mouse model of psoriasis at 5 mg/kg
Catalog No. 50-294-4586 Supplier Cayman Chemical Supplier No. 481000500 DTN
A steroidal glycoside with diverse biological activities reduces the proliferation of LNCaP and T47D cancer cells (IC50s 32.1 and 27.5 UM respectively) inhibits TNF-a-induced nuclear translocation of NF-KB in HaCaT keratinocytes at 50 nM decreases AST serum levels in a mouse model of carbon tetrachloride-induced liver injury at 0.1 mg/kg topical administration reduces ear epidermal thickness and scaling as well as leukocyte and macrophage infiltration in an imiquimod-induced mouse model of psoriasis at 5 mg/kg