Learn More
Medchemexpress LLC Cyclic AMP (sodium) | 37839-81-9 | 100.0% | 10 MM, 1 ML
SDP
Catalog No. 5000411359
Encompass_Preferred
Cyclic AMP (Cyclic adenosine monophosphate) sodium, an adenosine triphosphate derivative, is an intracellular signaling molecule that directs cellular responses to extracellular signals. It serves as an important second messenger in many biological processes.
  • Intracellular signaling molecule
  • Directs cellular responses to extracellular signals
  • Important second messenger in many biological processes
  • Modulates mediator generation
  • Suppresses the expression of pro-inflammatory cytokines (TNF-α and IL-12)
  • Enhances the production of the anti-inflammatory cytokine IL-10
Catalog No. 50-004-11359 Supplier Medchemexpress LLC Supplier No. HYB1511A10MM/1ML
Cyclic AMP (Cyclic adenosine monophosphate) sodium, an adenosine triphosphate derivative, is an intracellular signaling molecule that directs cellular responses to extracellular signals. It serves as an important second messenger in many biological processes.
  • Intracellular signaling molecule
  • Directs cellular responses to extracellular signals
  • Important second messenger in many biological processes
  • Modulates mediator generation
  • Suppresses the expression of pro-inflammatory cytokines (TNF-α and IL-12)
  • Enhances the production of the anti-inflammatory cytokine IL-10