Learn More
Medchemexpress LLC Cyclic AMP sodium | 37839-81-9 | 99.97% | 250 MG
SDP
Catalog No. 5000414491
Encompass_Preferred
Cyclic AMP (Cyclic adenosine monophosphate) sodium, an adenosine triphosphate derivative, is an intracellular signaling molecule that directs cellular responses to extracellular signals. It acts as an important second messenger in many biological processes. It also modulates mediator generation, suppressing the expression of pro-inflammatory cytokines and enhancing the production of anti-inflammatory cytokine.
  • Acts as an important second messenger
  • Modulates mediator generation
  • Suppresses pro-inflammatory cytokines like TNF-α and IL-12
  • Enhances anti-inflammatory cytokine IL-10 production
Catalog No. 50-004-14491 Supplier Medchemexpress LLC Supplier No. 250MGHYB1511A
Cyclic AMP (Cyclic adenosine monophosphate) sodium, an adenosine triphosphate derivative, is an intracellular signaling molecule that directs cellular responses to extracellular signals. It acts as an important second messenger in many biological processes. It also modulates mediator generation, suppressing the expression of pro-inflammatory cytokines and enhancing the production of anti-inflammatory cytokine.
  • Acts as an important second messenger
  • Modulates mediator generation
  • Suppresses pro-inflammatory cytokines like TNF-α and IL-12
  • Enhances anti-inflammatory cytokine IL-10 production