Learn More
Cayman Chemical CyclIc AMP TR-FRET KIt 1ea
SDP
Catalog No. 502994953
Encompass_Preferred
Designed to allow the user to detect and quantitatively measure the amount of cyclic AMP (cAMP) present in cell based assay systems.
Catalog No. 50-299-4953 Supplier Cayman Chemical Supplier No. 230421 EA
Designed to allow the user to detect and quantitatively measure the amount of cyclic AMP (cAMP) present in cell based assay systems.