Learn More
Invivogen c-di-AMP, 1 mg
SDP
Catalog No. NC0432587
Encompass
Bis-(3'-5')-cyclic dimeric adenosine monophosphate (c-di-AMP) is a bacterial second messenger implicated in the control of cell wall metabolism, osmotic stress responses and sporulation. Detection of c-di-AMP by the host cytoplasmic surveillance pathway (CSP) is known to elicit type I IFN responses through a signaling axis that involves STING, TBK1 and IRF3.
Catalog No. NC0432587 Supplier Invivogen Supplier No. TLRLNACDA
Bis-(3'-5')-cyclic dimeric adenosine monophosphate (c-di-AMP) is a bacterial second messenger implicated in the control of cell wall metabolism, osmotic stress responses and sporulation. Detection of c-di-AMP by the host cytoplasmic surveillance pathway (CSP) is known to elicit type I IFN responses through a signaling axis that involves STING, TBK1 and IRF3.