Learn More
Apexbio Technology LLC Cyclic di-GMP(Synonyms: Cyclic diguanylate, c-di-GMP, diguanylate cyclase product), 1mg, CAS: 61093-23-0.
SDP
Catalog No. 5000060237
Encompass_Preferred
Cyclic di-GMP (CAS 61093-23-0) is an intracellular second messenger involved in diverse bacterial physiological processes including biofilm formation motility and pathogenicity It functions in mammalian cells as an agonist for Stimulator of Interferon Genes (STING) directly binding and activating it to induce innate immune responses Due to this mechanism cyclic di-GMP serves as a valuable tool for investigating STING-mediated immune modulation pathways and has research applications in cancer immunotherapy studies notably enhancing antitumor immunity in metastatic melanoma models
Catalog No. 50-000-60237 Supplier Apexbio Technology LLC Supplier No. B78391
Cyclic di-GMP (CAS 61093-23-0) is an intracellular second messenger involved in diverse bacterial physiological processes including biofilm formation motility and pathogenicity It functions in mammalian cells as an agonist for Stimulator of Interferon Genes (STING) directly binding and activating it to induce innate immune responses Due to this mechanism cyclic di-GMP serves as a valuable tool for investigating STING-mediated immune modulation pathways and has research applications in cancer immunotherapy studies notably enhancing antitumor immunity in metastatic melanoma models