Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclic-di-GMP diammonium | 609343-82-0 | MFCD32708541 | 99.1% | 724.47 | C20H30N12O14P2 | 10 MG
SDP

Catalog No. 5000240829
Encompass_Preferred

Cyclic-di-GMP diammonium is a cyclic dinucleotide research reagent that functions as a STING agonist and a bacterial second messenger. It coordinates bacterial behaviors such as motility, virulence, biofilm formation, and cell cycle progression, and has reported activity in cellular assays including effects on cell proliferation and CD4 receptor expression. Supplied as the diammonium salt for laboratory research.

  • Used for STING activation and bacterial second-messenger studies.
  • Shows activity in cell proliferation and CD4 receptor expression assays.
  • Supplied as a diammonium salt; chemical formula C20H30N12O14P2.
  • Molecular weight 724.47.
  • Purity 99.07%.
  • Available in small research quantities suitable for in vitro experiments.

Catalog No. 50-002-40829 Supplier Medchemexpress LLC Supplier No. HY107780B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclic-di-GMP diammonium is a cyclic dinucleotide research reagent that functions as a STING agonist and a bacterial second messenger. It coordinates bacterial behaviors such as motility, virulence, biofilm formation, and cell cycle progression, and has reported activity in cellular assays including effects on cell proliferation and CD4 receptor expression. Supplied as the diammonium salt for laboratory research.

  • Used for STING activation and bacterial second-messenger studies.
  • Shows activity in cell proliferation and CD4 receptor expression assays.
  • Supplied as a diammonium salt; chemical formula C20H30N12O14P2.
  • Molecular weight 724.47.
  • Purity 99.07%.
  • Available in small research quantities suitable for in vitro experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.