Learn More
TARGETMOL CHEMICALS INC Cyclic-di-GMP disodium 1MG
SDP
Catalog No. 502876740
Encompass_Preferred
Also available in 5 mg, 10 mg and bulk. Please contact Fisher for quotes. Cyclic-di-GMP disodium (5GP-5GP disodium) is a STING agonist and a second messenger in bacteria. It is involved in a variety of processes in prokaryotes, including biofilm formation, motility, and cell cycle progression. Cyclic-di-GMP disodium (5GP-5GP disodium) exhibits antiproliferative activity against cancer cells, inducing increased CD4 receptor expression and cell cycle arrest, and is a novel therapeutic agent that can be developed to prevent or treat cancer. Purity 99.93%
Catalog No. 50-287-6740 Supplier TARGETMOL CHEMICALS INC Supplier No. T369831MG
Also available in 5 mg, 10 mg and bulk. Please contact Fisher for quotes. Cyclic-di-GMP disodium (5GP-5GP disodium) is a STING agonist and a second messenger in bacteria. It is involved in a variety of processes in prokaryotes, including biofilm formation, motility, and cell cycle progression. Cyclic-di-GMP disodium (5GP-5GP disodium) exhibits antiproliferative activity against cancer cells, inducing increased CD4 receptor expression and cell cycle arrest, and is a novel therapeutic agent that can be developed to prevent or treat cancer. Purity 99.93%