Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclic-di-GMP disodium | 2222132-40-1 | 98.78% | 734.37 | 100 MG
SDP

Catalog No. 5000343296
Encompass_Preferred

Cyclic-di-GMP disodium is a STING agonist and a bacterial second messenger that coordinates various aspects of bacterial growth and behavior, including motility, virulence, biofilm formation, and cell cycle progression. It exhibits anti-cancer cell proliferation activity and also induces elevated CD4 receptor expression and cell cycle arrest. Cyclic-di-GMP disodium can be utilized in cancer research.

  • Purity: 98.78%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Initial source: Endogenous metabolite
  • Shipping: Shipped with dry ice
  • Storage: -80°C, protect from light, stored under nitrogen

Catalog No. 50-003-43296 Supplier Medchemexpress LLC Supplier No. HY1103821MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cyclic-di-GMP disodium is a STING agonist and a bacterial second messenger that coordinates various aspects of bacterial growth and behavior, including motility, virulence, biofilm formation, and cell cycle progression. It exhibits anti-cancer cell proliferation activity and also induces elevated CD4 receptor expression and cell cycle arrest. Cyclic-di-GMP disodium can be utilized in cancer research.

  • Purity: 98.78%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Initial source: Endogenous metabolite
  • Shipping: Shipped with dry ice
  • Storage: -80°C, protect from light, stored under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.