Learn More
Cayman Chemical CyclIc dI-UMPsodIum salt 600ug
SDP
Catalog No. 502948176
Encompass_Preferred
A pyrimidine-containing CDN; binds to cGAS, in the apo or dsDNA-bound forms, with reduced affinity compared to 2’3’-cGAMP or 3’3’-cGAMP; intended for use as a negative control for cyclic di-GMP signaling
Catalog No. 50-294-8176 Supplier Cayman Chemical Supplier No. 33890600 UG
A pyrimidine-containing CDN; binds to cGAS, in the apo or dsDNA-bound forms, with reduced affinity compared to 2’3’-cGAMP or 3’3’-cGAMP; intended for use as a negative control for cyclic di-GMP signaling