Learn More
Medchemexpress LLC Cyclic diadenylate, diammonium salt | 00-00-0 | 658.4 g/mol | C20H24N10O12P2 | 5 MG
SDP
Catalog No. 5000264378
Encompass_Preferred
Cyclic diadenylate (c-di-AMP) diammonium is a cyclic dinucleotide that functions as a bacterial second messenger and a STING agonist, activating the TBK3-IRF3 signaling pathway to induce type I interferons and other cytokines. It is used in immunology research as an innate immune modulator and mucosal adjuvant, supplied as the diammonium salt in small research quantities.
  • Acts as a STING agonist to activate TBK3-IRF3 signaling.
  • Modulates innate immune responses and serves as a mucosal adjuvant.
  • Diammonium salt form suitable for research use.
  • Applicable to in vitro and in vivo immunology studies.
  • Supplied in small research quantities (for example, 5 mg).
Catalog No. 50-002-64378 Supplier Medchemexpress LLC Supplier No. HY12326B5MG
Cyclic diadenylate (c-di-AMP) diammonium is a cyclic dinucleotide that functions as a bacterial second messenger and a STING agonist, activating the TBK3-IRF3 signaling pathway to induce type I interferons and other cytokines. It is used in immunology research as an innate immune modulator and mucosal adjuvant, supplied as the diammonium salt in small research quantities.
  • Acts as a STING agonist to activate TBK3-IRF3 signaling.
  • Modulates innate immune responses and serves as a mucosal adjuvant.
  • Diammonium salt form suitable for research use.
  • Applicable to in vitro and in vivo immunology studies.
  • Supplied in small research quantities (for example, 5 mg).