Learn More
Medchemexpress LLC Cyclic diadenylate disodium | 2734909-87-4 | 99.1% | 702.38 | C20H22N10Na2O12P2 | 10MM 1ML
SDP
Catalog No. 5000235429
Encompass_Preferred
Cyclic diadenylate disodium (c-di-AMP disodium) is the disodium salt of cyclic diadenylate supplied as a 10 mM solution in DMSO (1 mL). It functions as a bacterial second messenger and a STING agonist used in innate immunity, adjuvant research, and bacterial signaling studies. Handle as a research reagent, store at -80°C, and protect from light.
Catalog No. 50-002-35429 Supplier Medchemexpress LLC Supplier No. HY12326A10MM 1ML
Cyclic diadenylate disodium (c-di-AMP disodium) is the disodium salt of cyclic diadenylate supplied as a 10 mM solution in DMSO (1 mL). It functions as a bacterial second messenger and a STING agonist used in innate immunity, adjuvant research, and bacterial signaling studies. Handle as a research reagent, store at -80°C, and protect from light.