Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclic diadenylate disodium | 2734909-87-4 | 99.1% | 702.38 g/mol | C20H22N10Na2O12P2 | 50MG
SDP

Catalog No. 5000235427
Encompass_Preferred

c-di-AMP disodium is the disodium salt of cyclic diadenylate, a bacterial second messenger and STING agonist used to activate the TBK1-IRF3 pathway and study innate immune signaling in vitro. It is supplied as a white to off-white solid and is provided in multiple solid and solution sizes for research and analytical applications.

  • Acts as a STING agonist for innate immunity and signaling studies.
  • High purity for research use (99.1%).
  • Soluble in DMSO and water; DMSO up to 270 mg/mL, water ≥50 mg/mL.
  • Molecular weight 702.38 g/mol; formula C20H22N10Na2O12P2.
  • Store at -80°C, protect from light; stock solutions stable up to 6 months under nitrogen.

Catalog No. 50-002-35427 Supplier Medchemexpress LLC Supplier No. HY12326A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

c-di-AMP disodium is the disodium salt of cyclic diadenylate, a bacterial second messenger and STING agonist used to activate the TBK1-IRF3 pathway and study innate immune signaling in vitro. It is supplied as a white to off-white solid and is provided in multiple solid and solution sizes for research and analytical applications.

  • Acts as a STING agonist for innate immunity and signaling studies.
  • High purity for research use (99.1%).
  • Soluble in DMSO and water; DMSO up to 270 mg/mL, water ≥50 mg/mL.
  • Molecular weight 702.38 g/mol; formula C20H22N10Na2O12P2.
  • Store at -80°C, protect from light; stock solutions stable up to 6 months under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.