Learn More
Cayman Chemical CyclIc dIAMPHRP Tracer 100dtn
SDP
Catalog No. 502942358
Encompass_Preferred
Cyclic di-AMP-HRP Tracer has been tested and formulated to work exclusively with Cayman’s Cyclic di-AMP ELISA Kit
Catalog No. 50-294-2358 Supplier Cayman Chemical Supplier No. 401960100 DTN
Cyclic di-AMP-HRP Tracer has been tested and formulated to work exclusively with Cayman’s Cyclic di-AMP ELISA Kit