Learn More
Cayman Chemical CyclIc dIGMPHRP Tracer 100dtn
SDP
Catalog No. 502942298
Encompass_Preferred
Cyclic di-GMP-HRP Tracer has been tested and formulated to work exclusively with Cayman’s Cyclic di-GMP ELISA Kit (Item No. 501780). Please visit Cyclic di-GMP ELISA Kit (Item No. 501780) for the kit protocol, procedures, and product handling.
Catalog No. 50-294-2298 Supplier Cayman Chemical Supplier No. 401780100 DTN
Cyclic di-GMP-HRP Tracer has been tested and formulated to work exclusively with Cayman’s Cyclic di-GMP ELISA Kit (Item No. 501780). Please visit Cyclic di-GMP ELISA Kit (Item No. 501780) for the kit protocol, procedures, and product handling.