Learn More
Medchemexpress LLC Cyclic GMP | 7665-99-8 | 99.8% | 345.21 | 25 MG
SDP
Catalog No. 5000438595
Encompass_Preferred
Cyclic GMP (cGMP) is an important second messenger and a major intracellular mediator of extracellular signals such as nitric oxide (NO) and natriuretic peptides (NPs). Its effects are primarily through cGMP-dependent protein kinases (PKGs), cGMP-gated cation channels, and phosphodiesterases (PDEs). Cyclic GMP can also inhibit platelet adhesion and aggregation. cGAMP (Cyclic-GMP-AMP), a conjugate of Cyclic GMP and AMP, can induce IRF3 phosphorylation and nuclear translocation, thereby enhancing antiviral immune responses.
  • Important second messenger.
  • Major intracellular mediator of extracellular signals.
  • Acts primarily through cGMP-dependent protein kinases, cGMP-gated cation channels, and phosphodiesterases.
  • Can inhibit platelet adhesion and aggregation.
  • Conjugate cGAMP can induce IRF3 phosphorylation and nuclear translocation.
  • Enhances antiviral immune responses.
Catalog No. 50-004-38595 Supplier Medchemexpress LLC Supplier No. 25MGHY113469
Cyclic GMP (cGMP) is an important second messenger and a major intracellular mediator of extracellular signals such as nitric oxide (NO) and natriuretic peptides (NPs). Its effects are primarily through cGMP-dependent protein kinases (PKGs), cGMP-gated cation channels, and phosphodiesterases (PDEs). Cyclic GMP can also inhibit platelet adhesion and aggregation. cGAMP (Cyclic-GMP-AMP), a conjugate of Cyclic GMP and AMP, can induce IRF3 phosphorylation and nuclear translocation, thereby enhancing antiviral immune responses.
  • Important second messenger.
  • Major intracellular mediator of extracellular signals.
  • Acts primarily through cGMP-dependent protein kinases, cGMP-gated cation channels, and phosphodiesterases.
  • Can inhibit platelet adhesion and aggregation.
  • Conjugate cGAMP can induce IRF3 phosphorylation and nuclear translocation.
  • Enhances antiviral immune responses.