Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cyclic GMP-AMP diammonium | 96.6% | 708.47 g/mol | C20H30N12O13P2 | 5 MG
SDP

Catalog No. 5000263475
Encompass_Preferred

cGAMP diammonium is a cyclic dinucleotide that functions as an endogenous second messenger and a potent activator of the stimulator of interferon genes (STING) pathway. Provided as the diammonium salt for research use, it induces type I interferon production and is used to study innate immune signaling.

  • Activates STING and triggers type I interferon signaling.
  • High purity (96.6%) suitable for research applications.
  • Molecular formula C20H30N12O13P2; molecular weight 708.47 g/mol.
  • Water-soluble; dissolves up to 100 mg/mL with ultrasonic assistance.
  • Solid storage recommended at -20°C; solution storage guidance provided.
  • Supplied in small quantities (e.g., 5 mg) for laboratory use.

Catalog No. 50-002-63475 Supplier Medchemexpress LLC Supplier No. HY110385A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cGAMP diammonium is a cyclic dinucleotide that functions as an endogenous second messenger and a potent activator of the stimulator of interferon genes (STING) pathway. Provided as the diammonium salt for research use, it induces type I interferon production and is used to study innate immune signaling.

  • Activates STING and triggers type I interferon signaling.
  • High purity (96.6%) suitable for research applications.
  • Molecular formula C20H30N12O13P2; molecular weight 708.47 g/mol.
  • Water-soluble; dissolves up to 100 mg/mL with ultrasonic assistance.
  • Solid storage recommended at -20°C; solution storage guidance provided.
  • Supplied in small quantities (e.g., 5 mg) for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.